Learn More
New England Biolab Ltd BstXI 5000 units
Numéro de catalogue. 50811948
Encompass
A restriction endonuclease that recognizes the sequence CCAN_NNNN^NTGG.
Numéro de catalogue. 50811948 Fournisseur New England Biolab Ltd Code fournisseur R0113L
A restriction endonuclease that recognizes the sequence CCAN_NNNN^NTGG.