Learn More
New England Biolab Ltd BssSI-v2 200 units
Numéro de catalogue. NC0836942
Encompass
BssSI-v2 is an engineered and improved version of BssSI that recognizes the sequence VC^TCGAGB. BssSI-v2 is an engineered and improved version of BssSI. (NEB uses the designation "v2" or "HF" in the name to indicate engineered enzymes.)
Numéro de catalogue. NC0836942 Fournisseur New England Biolab Ltd Code fournisseur R0680S
Pour de l'aide SVP utiliser la function de clavardage; nous envoyer un couriel à help@thermofisher.com ou appelez le service à la clientèle au 1-800-234-7437.
BssSI-v2 is an engineered and improved version of BssSI that recognizes the sequence VC^TCGAGB. BssSI-v2 is an engineered and improved version of BssSI. (NEB uses the designation "v2" or "HF" in the name to indicate engineered enzymes.)