missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Bromothymol Blue TS, 0.1% (w/v) in 50% (v/v) Alcohol, Ricca Chemical
$294.87 - $294.87
Chemical Identifiers
| CAS | 76-59-5 |
|---|---|
| Molecular Formula | C27H28Br2O5S |
| Molecular Weight (g/mol) | Mixture |
| InChI Key | NUHCTOLBWMJMLX-UHFFFAOYSA-N |
| PubChem CID | 6450 |
| ChEBI | CHEBI:86155 |
Specifications
| 76-59-5 , 64-17-5 , 67-56-1 , 7732-18-5 | |
| C27H28Br2O5S | |
| CC1=C(C(=C(C=C1C2(C3=CC=CC=C3S(=O)(=O)O2)C4=CC(=C(C(=C4C)Br)O)C(C)C)C(C)C)O)Br | |
| Mixture | |
| CHEBI:86155 | |
| Natural Poly Bottle | |
| Orange/Red | |
| Liquid |
| 0.1% (w/v) | |
| NUHCTOLBWMJMLX-UHFFFAOYSA-N | |
| 2-bromo-4-[3-(3-bromo-4-hydroxy-2-methyl-5-propan-2-ylphenyl)-1,1-dioxo-2,1$l^{6}-benzoxathiol-3-yl]-3-methyl-6-propan-2-ylphenol | |
| 6450 | |
| Indicator | |
| 50% (v/v) Alcohol | |
| 120 mL | |
| Bromothymol Blue, Test Solution |