missing translation for 'onlineSavingsMsg'
Learn More

Bromothymol Blue Sodium Salt, TCI America™

Catalog No. S00445G
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S00445G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S00445G Supplier TCI America Supplier No. S00445G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 34722-90-2
Molecular Formula C27H27Br2NaO5S
Molecular Weight (g/mol) 646.37
MDL Number MFCD00077263,MFCD00077263,MFCD00077263
InChI Key NMKFVGALBGZKGW-FKWCIMQXSA-M
Synonym cfn0fbc3yh, unii-cfn0fbc3yh, sodium bromothymol blue, bromothymol blue sodium salt, 3',3-dibromothymolsulfonephthalein sodium salt, bromthymol blue sodium, phenol, 4,4'-1,1-dioxido-3h-2,1-benzoxathiol-3-ylidene bis 2-bromo-3-methyl-6-1-methylethyl-, monosodium salt, phenol, 4,4'-1,1-dioxido-3h-2,1-benzoxathiol-3-ylidene bis 2-bromo-3-methyl-6-1-methylethyl-, sodium salt 1:1, bromthymol blue sodium salt, bromothymol blue, sodium salt
PubChem CID 102183223
IUPAC Name sodium 2-{[(1Z)-3-bromo-2-methyl-4-oxo-5-(propan-2-yl)cyclohexa-2,5-dien-1-ylidene][3-bromo-4-hydroxy-2-methyl-5-(propan-2-yl)phenyl]methyl}benzene-1-sulfonate
SMILES [Na+].CC(C)C1=CC(\C(C2=CC=CC=C2S([O-])(=O)=O)=C2/C=C(C(C)C)C(=O)C(Br)=C2C)=C(C)C(Br)=C1O

Specifications

Quantity 5 g
Formula Weight 646.37
Physical Form Crystalline Powder
Chemical Name or Material Bromothymol Blue Sodium Salt