missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Bromothymol Blue sodium salt

Catalog No. AAA1791318
Change view
Click to view available options
Quantity:
10 g
50 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1791318 50 g
AAA1791309 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1791318 Supplier Thermo Scientific Chemicals Supplier No. A1791318
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 34722-90-2 | C27H27Br2NaO5S | 646.37 g/mol

It is employed as a pH indicator which is yellow at pH ? 6.0 and blue at pH ? 7.6. It finds its major application in lithium battery, textiles and antagonists.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is employed as a pH indicator which is yellow at pH ≤ 6.0 and blue at pH ≥ 7.6. It finds its major application in lithium battery, textiles and antagonists.

Solubility
Soluble in water and ethanol.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.

Chemical Identifiers

CAS 34722-90-2
Molecular Formula C27H27Br2NaO5S
Molecular Weight (g/mol) 646.37
MDL Number MFCD00077263,MFCD00077263,MFCD00077263
InChI Key NMKFVGALBGZKGW-FKWCIMQXSA-M
Synonym Bromothymol Blue, water soluble
PubChem CID 102183223
IUPAC Name sodium;2-bromo-4-[(Z)-(3-bromo-2-methyl-4-oxo-5-propan-2-ylcyclohexa-2,5-dien-1-ylidene)-(2-sulfophenyl)methyl]-3-methyl-6-propan-2-ylphenolate
SMILES [Na+].CC(C)C1=CC(\C(C2=CC=CC=C2S([O-])(=O)=O)=C2/C=C(C(C)C)C(=O)C(Br)=C2C)=C(C)C(Br)=C1O

Specifications

Melting Point 200°C to 202°C
Solubility Information Soluble in water and ethanol.
Quantity 50 g
Formula Weight 646.38
Chemical Name or Material Bromothymol Blue sodium salt
EINECSNumber 252-169-7
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only