missing translation for 'onlineSavingsMsg'
Learn More

Bromothymol Blue Absorbance Standard, 23.2 μM in dilute buffer pH 8.0, Ricca Chemical

Catalog No. 136925
Change view
Click to view available options
Quantity:
10 L
Packaging:
Cubitainer
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
136925 10 L Cubitainer
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 136925 Supplier Ricca Chemical Company Supplier No. R136900010F

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Specifications

Chemical Name or Material Bromothymol Blue Absorbance Standard
Name Note 23.2μM in dilute buffer pH 8.0
CAS 34722-90-2, 7732-18-5, 7778-77-0, 1310-73-2
Odor Odorless
Packaging Cubitainer
Solvent or Matrix Aqueous Solution
Color Blue
Solubility Information Solubility in Water: Infinite
Quantity 10 L
Grade Indicator