missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Bromothymol Blue, 0.04% Aqueous Solution

Catalog No. AC612405000
Change view
Click to view available options
Quantity:
500 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC612405000 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC612405000 Supplier Thermo Scientific Chemicals Supplier No. 612405000
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

 

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Indicator

EcoSafPak™ is an environmentally friendly packaging system made of 100% recyclable material by a SFI certified supplier.

Chemical Identifiers

CAS 34722-90-2
Molecular Formula C27H27Br2NaO5S
Molecular Weight (g/mol) 646.37
MDL Number MFCD00077263,MFCD00077263,MFCD00077263
InChI Key NMKFVGALBGZKGW-FKWCIMQXSA-M
Synonym Bromthymol Blue, sodium salt, 3', 3''-Dibromothymolsulfonephthalein, sodium salt, BTB
PubChem CID 102183223
IUPAC Name sodium 2-{[(1Z)-3-bromo-2-methyl-4-oxo-5-(propan-2-yl)cyclohexa-2,5-dien-1-ylidene][3-bromo-4-hydroxy-2-methyl-5-(propan-2-yl)phenyl]methyl}benzene-1-sulfonate
SMILES [Na+].CC(C)C1=CC(\C(C2=CC=CC=C2S([O-])(=O)=O)=C2/C=C(C(C)C)C(=O)C(Br)=C2C)=C(C)C(Br)=C1O

Specifications

Concentration 0.04%
Packaging EcoSafPak™
Visual Transition Interval Pass Test
Merck Index 15, 1455
pH 6.5 to 7.1
Quantity 500 mL
Formula Weight 646.37
Chemical Name or Material Bromothymol Blue

RUO – Research Use Only