missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Bromothymol Blue, 0.04% Aqueous, pH 6.0 to 7.6 Yellow to Blue, Certified, LabChem™
Specifications
Specifications
| CAS Max % | 99.96, 0.04 |
| Identification | Passes Test |
| Packaging | Poly Bottle |
| Specific Gravity | 1g/mL |
| Linear Formula | C27H27Br2O5S·Na |
| Decomposition Information | Nitrogen oxides; Carbon monoxide; Carbon dioxide |
| Density | 1g/mL |
| Color | Green |
| Solubility Information | Soluble in water |
| Quantity | 125 mL |
| Show More |
Safety and Handling
| Hazard Statement | GHS H Statement Solution is not hazardous. |
| Precautionary Statement | GHS P Statement If in contact with skin or eyes, rinse thoroughly with water for 15-20 minutes. If swallowed, get medical attention. |
| Recommended Storage | Room Temperature |