missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Bromophenol Blue Indicator, 0.4% (w/v), Ricca Chemical
$337.33 - $337.33
Specifications
| Chemical Name or Material | Bromophenol Blue Indicator |
|---|---|
| CAS | 62625-28-9 , 7732-18-5 |
| CAS Min % | 98.0,0.39 |
| CAS Max % | 100.0,0.41 |
| Concentration | 0.4% (w/v) |
Specifications
| Bromophenol Blue Indicator | |
| 98.0,0.39 | |
| 0.4% (w/v) | |
| Indicator | |
| Aqueous Solution | |
| 4 |
| 62625-28-9 , 7732-18-5 | |
| 100.0,0.41 | |
| 0.4% W/V | |
| Natural Poly Bottle | |
| Dark Reddish Purple | |
| Liquid |