missing translation for 'onlineSavingsMsg'
Learn More

Bromomethyl Methyl Ether 95.0+%, TCI America™

Catalog No. B21315G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B21315G 5 g
B213125G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B21315G Supplier TCI America Supplier No. B21315G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 13057-17-5
Molecular Formula C2H5BrO
Molecular Weight (g/mol) 124.965
MDL Number MFCD00000171
InChI Key JAMFGQBENKSWOF-UHFFFAOYSA-N
Synonym bromomethyl methyl ether, methane, bromomethoxy, bromo methoxy methane, methoxymethyl bromide, ether, bromomethyl methyl, bromomethylmethylether, methoxymethylbromide, mombr, bromo-methoxy-methane, bromomethyl methylether
PubChem CID 83093
IUPAC Name bromo(methoxy)methane
SMILES COCBr

Specifications

Color Yellow
Boiling Point 88°C
Quantity 5 g
UN Number 1992
Formula Weight 124.97
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material Bromomethyl Methyl Ether
TSCA Yes
Recommended Storage Refrigerator