missing translation for 'onlineSavingsMsg'
Learn More

(Bromomethyl)cyclopropane, 96%, Thermo Scientific™

Numéro de catalogue. AC221810250
Modifier l'affichage
Click to view available options
Quantité:
1g
5g
25g
Emballage:
Glass bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantité Emballage
AC221810250 25g Glass bottle
AC221810010 1g Glass bottle
AC221810050 5g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Numéro de catalogue. AC221810250 Fournisseur Thermo Scientific Chemicals Code fournisseur 221810250

Pour de l'aide SVP utiliser la function de clavardage; nous envoyer un couriel à help@thermofisher.com ou appelez le service à la clientèle au 1-800-234-7437.

Pour de l'aide SVP utiliser la function de clavardage; nous envoyer un couriel à help@thermofisher.com ou appelez le service à la clientèle au 1-800-234-7437.

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Identifiants chimiques

CAS 7051-34-5
Formule moléculaire C4H7Br
Poids moléculaire (g/mol) 135.00
Numéro MDL MFCD00001306
Clé InChI AEILLAXRDHDKDY-UHFFFAOYSA-N
Synonyme bromomethyl cyclopropane, cyclopropylmethyl bromide, cyclopropane, bromomethyl, cyclopropyl bromo methane, 1-bromomethyl cyclopropane, bromocyclopropylmethane, cyclopropylmethylbromide, bromomethyl-cyclopropane, cyclopropyl methylbromide
PubChem CID 81503
Nom de l’IUPAC bromomethylcyclopropane
SOURIRES BrCC1CC1

Spécifications

Nom ou substance chimique (Bromomethyl)cyclopropane
CAS % minimum 95.0
CAS % maximal 100.0
Densité 1.3800g/mL
Point d’ébullition 101.0°C to 108.0°C
Point d’éclair 41°C
Spectre infrarouge Authentic
Fourchette de pourcentages d’essai 95% min. (GC)
Emballage Glass bottle
Indice de réfraction 1.4750 to 1.4790
Formule linéaire C3H5CH2Br
Quantité 25g
Gravité spécifique 1.38
Poids de formule 135
Pourcentage de pureté 96%
Afficher plus de résultats Afficher moins de résultats
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Causes skin irritation.
Harmful if swallowed.
Causes serious eye irritation.
May cause respiratory irritation.
Flammable liquid and vapor.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 230-331-8

RUO – Research Use Only