missing translation for 'onlineSavingsMsg'
Learn More

Bromohydroquinone 90.0+%, TCI America™

Supplier:  TCI America B16005G

 View more versions of this product

Catalog No. B16005G

Only null left
Explore available promotions

Description

Description

Specifications

Specifications

Bromohydroquinone
C6H5BrO2
5 g
REFDOIWRJDGBHY-UHFFFAOYSA-N
2-bromobenzene-1,4-diol
68502
≥90.0% (GC)
583-69-7
MFCD00041747
bromohydroquinone, 2-bromohydroquinone, 1,4-benzenediol, 2-bromo, 2-bromoquinol, 2-bromo-1,4-benzenediol, hydroquinone, bromo, bromoquinol, 4-hydroxy bromophenol, 2-bromo-benzene-1,4-diol
C1=CC(=C(C=C1O)Br)O
189.008
189.01
Crystalline Powder

Chemical Identifiers

583-69-7
189.008
REFDOIWRJDGBHY-UHFFFAOYSA-N
68502
C1=CC(=C(C=C1O)Br)O
C6H5BrO2
MFCD00041747
bromohydroquinone, 2-bromohydroquinone, 1,4-benzenediol, 2-bromo, 2-bromoquinol, 2-bromo-1,4-benzenediol, hydroquinone, bromo, bromoquinol, 4-hydroxy bromophenol, 2-bromo-benzene-1,4-diol
2-bromobenzene-1,4-diol
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

RTECSNumber : CZ8920000

TSCA : Yes

SDS
Documents

Documents

Promotions

Promotions