missing translation for 'onlineSavingsMsg'
Learn More

Bromohydroquinone 90.0+%, TCI America™

Catalog No. p-7119145
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B16005G 5 g
B160025G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B16005G Supplier TCI America Supplier No. B16005G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 583-69-7
Molecular Formula C6H5BrO2
Molecular Weight (g/mol) 189.008
MDL Number MFCD00041747
InChI Key REFDOIWRJDGBHY-UHFFFAOYSA-N
Synonym bromohydroquinone, 2-bromohydroquinone, 1,4-benzenediol, 2-bromo, 2-bromoquinol, 2-bromo-1,4-benzenediol, hydroquinone, bromo, bromoquinol, 4-hydroxy bromophenol, 2-bromo-benzene-1,4-diol
PubChem CID 68502
IUPAC Name 2-bromobenzene-1,4-diol
SMILES C1=CC(=C(C=C1O)Br)O

Specifications

Quantity 5 g
Formula Weight 189.01
Percent Purity ≥90.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Bromohydroquinone
RTECSNumber CZ8920000
TSCA Yes