accessibility menu, dialog, popup

UserName

CAS RN 583-69-7

IUPAC Name: 2-bromobenzene-1,4-diol
Synonyms: 1,4-benzenediol, 2-bromo-
Molecular Weight (g/mol): 189.01
Molecular Formula: C6H5BrO2
InChi Key: REFDOIWRJDGBHY-UHFFFAOYSA-N
SMILES: OC1=CC=C(O)C(Br)=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results