missing translation for 'onlineSavingsMsg'
Learn More

Bromodimethylsulfonium Bromide 98.0+%, TCI America™

Catalog No. B33115G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B33115G 5 g
B331125G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B33115G Supplier TCI America Supplier No. B33115G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 50450-21-0
Molecular Formula C2H6Br2S
Molecular Weight (g/mol) 221.94
MDL Number MFCD10000904
InChI Key RGVBVVVFSXWUIM-UHFFFAOYSA-M
PubChem CID 11481441
IUPAC Name bromodimethylsulfanium bromide
SMILES [Br-].C[S+](C)Br

Specifications

Melting Point 82°C
Quantity 5 g
UN Number 3261
Formula Weight 221.94
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Bromodimethylsulfonium Bromide
TSCA No
Recommended Storage Freezer