missing translation for 'onlineSavingsMsg'
Learn More

Bromocresol Purple 97.0+%, TCI America™

Catalog No. B058025G
Change view
Click to view available options
Quantity:
1 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B058025G 25 g
B05801G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B058025G Supplier TCI America Supplier No. B058025G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 115-40-2 | C21H16Br2O5S | 540.22 g/mol

Chemical Identifiers

CAS 115-40-2
Molecular Formula C21H16Br2O5S
Molecular Weight (g/mol) 540.22
MDL Number MFCD00011681
InChI Key ABIUHPWEYMSGSR-UHFFFAOYSA-N
PubChem CID 8273
ChEBI CHEBI:86154
IUPAC Name 3,3-bis(3-bromo-4-hydroxy-5-methylphenyl)-3H-2,1λ⁶-benzoxathiole-1,1-dione
SMILES CC1=CC(=CC(Br)=C1O)C1(OS(=O)(=O)C2=CC=CC=C12)C1=CC(C)=C(O)C(Br)=C1

Specifications

Melting Point 240°C
Color Orange
Quantity 25 g
Physical Form Crystal/Powder
Formula Weight 540.22
Percent Purity ≥97.0% (HPLC)
Grade Laboratory
Chemical Name or Material Bromocresol Purple
EINECSNumber (5)-0533
TSCA Yes