missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Bromocresol Purple (0.04% in Water) [for pH Determination], TCI America™
Specifications
Specifications
| Color | Red/Yellow |
| Quantity | 500 mL |
| Physical Form | Liquid |
| Formula Weight | 540.22 |
| Grade | Laboratory |
| Chemical Name or Material | Bromocresol Purple (0.04% in Water) [for pH Determination] |
Safety and Handling
| EINECSNumber | (5)-0533 |
| TSCA | Yes |