missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Bromocresol Green Sodium Salt, ACS Reagent Grade, Ricca Chemical
Specifications
Specifications
| CAS | 62625-32-5 |
| CAS Min % | 98.0 |
| CAS Max % | 100.0 |
| Grade | ACS Reagent |
| Packaging | White Poly Wide Mouth Bottle |
| Color | Green |
| Quantity | 200 g |
| Chemical Name or Material | Bromocresol Green, Sodium Salt |