missing translation for 'onlineSavingsMsg'
Learn More

Bromocresol Green Sodium Salt, ACS Reagent Grade, Ricca Chemical

Catalog No. RDCB060010F
Change view
Click to view available options
Quantity:
10 g
Packaging:
Pouch
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
RDCB060010F 10 g Pouch
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. RDCB060010F Supplier Ricca Chemical Company Supplier No. RDCB060010F1

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 62625-32-5 | 720 g/mol

Chemical Identifiers

CAS 62625-32-5
Molecular Weight (g/mol) 720 g/mol

Specifications

CAS Min % 98.0
CAS Max % 100.0
Packaging Pouch
Color Dark Green
Quantity 10 g
Physical Form Granules
Percent Purity ≥98%
Grade ACS
Chemical Name or Material Bromocresol Green, Sodium Salt