missing translation for 'onlineSavingsMsg'
Learn More

Bromocresol Green Sodium Salt, ACS Grade, LabChem™

Catalog No. LC118207
Change view
Click to view available options
Quantity:
10 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
LC118207 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. LC118207 Supplier LabChem Supplier No. LC118207

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

ACS grade pH indicator for chemical laboratory use.

Chemical Identifiers

CAS 62625-32-5
Molecular Weight (g/mol) 720.06

Specifications

Melting Point 230°C
CAS Max % 100
Identification Passes Test
Packaging Amber Glass
Linear Formula C21H14Br4O5S·Na
Decomposition Information Carbon monoxide; Carbon dioxide; Sulfur compounds; Hydrogen bromide
Color Gray
Solubility Information Soluble in water
Quantity 10 g
Physical Form Crystalline Powder
Formula Weight 720.06
Percent Purity 100%
Grade ACS
Chemical Name or Material Bromocresol Green Sodium Salt
Show More Show Less
Hazard Statement GHS H Statement
Product is not hazardous.
Precautionary Statement GHS P Statement
If in contact with skin or eyes, rinse thoroughly with water for 15-20 minutes.
If swallowed, get medical attention.
EINECSNumber 263-657-4
Recommended Storage Room Temperature