missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Bromocresol Green sodium salt, 0.04% w/v aq. soln.

Catalog No. AA38696AP
Change view
Click to view available options
Quantity:
100 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA38696AP 500 mL
AA38696AE 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA38696AP Supplier Thermo Scientific Chemicals Supplier No. 038696AP

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 62625-32-5

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 62625-32-5
MDL Number MFCD00148898
Synonym Bromocresol Green, water soluble

Specifications

Concentration 0.04% w/v aq. soln.
Color Green
Quantity 500 mL
Physical Form Liquid
Formula Weight 720.01
Chemical Name or Material Bromocresol Green sodium salt
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only