missing translation for 'onlineSavingsMsg'
Learn More

Bromocresol Green, 0.1% (w/v) Aqueous, pH 3.8 to 5.4 Yellow to Blue, Certified, LabChem™

Catalog No. LC118507
Change view
Click to view available options
Quantity:
125 mL
500 mL
Packaging:
Poly Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
LC118507 125 mL Poly Bottle
LC118501 500 mL Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. LC118507 Supplier LabChem Supplier No. LC118507

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 62625-32-5 | 720.06 g/mol

Chemical Identifiers

CAS 62625-32-5 , 7732-18-5

Specifications

CAS Max % 99.9, 0.1
Identification Passes Test
Packaging Poly Bottle
Specific Gravity 1g/mL
Linear Formula C21H14Br4O5S·Na
Decomposition Information Carbon monoxide; Carbon dioxide; Nitrogen oxides; Hydrogen bromide
Density 1g/mL
Color Green
pH 3.8 to 5.4
Solubility Information Soluble in water
Quantity 125 mL
Physical Form Liquid
Molecular Weight (g/mol) 720.06
Formula Weight 720.06
Percent Purity 0.1%
Grade Certified
Chemical Name or Material Bromocresol Green, 0.1% (w/v) Aqueous, pH 3.8 to 5.4 Yellow to Blue
Show More Show Less
Hazard Statement GHS H Statement
Solution is not hazardous.
Precautionary Statement GHS P Statement
If in contact with skin or eyes, rinse thoroughly with water for 15-20 minutes.
If swallowed, get medical attention.
Recommended Storage Room Temperature