missing translation for 'onlineSavingsMsg'
Learn More

Bromoacetic acid N-hydroxysuccinimide ester, 90%

Catalog No. AC427782500
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Packaging:
Glass bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC427782500 250 mg Glass bottle
AC427780010 1 g Glass bottle
AC427780050 5 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC427782500 Supplier Thermo Scientific Chemicals Supplier No. 427782500
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 42014-51-7 | C6H6BrNO4 | 236.02 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 42014-51-7
Molecular Formula C6H6BrNO4
Molecular Weight (g/mol) 236.02
MDL Number MFCD00058571
InChI Key NKUZQMZWTZAPSN-UHFFFAOYSA-N
Synonym n-succinimidyl bromoacetate, bromoacetic acid n-hydroxysuccinimide ester, 1-bromoacetyl oxy pyrrolidine-2,5-dione, 2,5-dioxoazolidinyl 2-bromoacetate, bromoacetic acid n-succinimidyl ester, pubchem11830, n-succinimidylbromoacetate, n-bromoacetoxy succinimide, bicl212, n-hydroxysuccinimide bromoacetate
PubChem CID 3565210
IUPAC Name (2,5-dioxopyrrolidin-1-yl) 2-bromoacetate
SMILES C1CC(=O)N(C1=O)OC(=O)CBr

Specifications

Color White
Elemental Analysis 88% min.
Infrared Spectrum Authentic
Assay Percent Range 90%
Packaging Glass bottle
Quantity 250 mg
Formula Weight 236.02
Percent Purity 90%
Physical Form Powder
Chemical Name or Material Bromoacetic acid N-hydroxysuccinimide ester
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

RUO – Research Use Only