missing translation for 'onlineSavingsMsg'
Learn More

Bromine Water, Saturated, approximately 3% (w/v), Ricca Chemical

Catalog No. 119532
Change view
Click to view available options
Quantity:
500 mL
1 L
Packaging:
Safety-coated Glass Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
119532 1 L Safety-coated Glass Bottle
119516 500 mL Safety-coated Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 119532 Supplier Ricca Chemical Company Supplier No. 119532

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 7726-95-6 , 7732-18-5
Molecular Formula Br
Molecular Weight (g/mol) 79.91
InChI Key CPELXLSAUQHCOX-UHFFFAOYSA-M
Synonym bromine, dibromine, brom, bromine solution, brome, bromo, broom, bromine water, bromo italian, bromo spanish
PubChem CID 24408
ChEBI CHEBI:29224
IUPAC Name bromide
SMILES [Br-]

Specifications

Color Brown-Red to Orange
CAS Min % 95.0, 2.95
CAS Max % 99.0, 3.07
Grade Laboratory
Packaging Safety-coated Glass Bottle
Quantity 1 L
Physical Form Liquid
Concentration or Composition Notes approximately 3% (w/v)
Chemical Name or Material Bromine Water, Saturated