missing translation for 'onlineSavingsMsg'
Learn More

Bromine liquid, 99.8%

Catalog No. AA0090514
Change view
Click to view available options
Quantity:
25 g
250 g
1 kg
5 x 1 kg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA0090514 25 g
AA0090530 250 g
AA00905A1 1 kg
AA00905D9 5 x 1 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. AA0090514 Supplier Thermo Scientific Chemicals Supplier No. 00090514

CAS: 7726-95-6 | Br | 79.91 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 7726-95-6
Molecular Formula Br
Molecular Weight (g/mol) 79.91
MDL Number MFCD00010896
InChI Key CPELXLSAUQHCOX-UHFFFAOYSA-M
Synonym bromine, dibromine, brom, bromine solution, brome, bromo, broom, bromine water, bromo italian, bromo spanish
PubChem CID 24408
ChEBI CHEBI:29224
SMILES [Br-]

Specifications

Physical Form Liquid
Quantity 25 g
Assay Percent Range 99.8%
IUPAC Name bromide
Formula Weight 159.81
Percent Purity 99.8%
Density 3.1g/cm3
Chemical Name or Material Bromine
Hazard Category H314-H330
Precautionary Statement P260-P264b-P271-P280-P284-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
DOTInformation Hazard Class: 8; Packaging Group: I
EINECSNumber 231-778-1
RTECSNumber EF9100000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only