missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Bromine, 99.5+%, ACS reagent
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Boiling Point | 58.7°C |
| Melting Point | -7.2°C |
| Quantity | 100 g |
| Packaging | Plastic Coated Glass Bottle |
| Specific Gravity | 3.111 |
| Assay Percent Range | 99.5% min. (Iodometry) |
| Density | 3.1110g/mL |
| Linear Formula | Br2 |
| Merck Index | 15,1401 |
| Solubility Information | Solubility in water: 35g/L (20°C). Other solubilities: soluble in concd hcl,aq solns of bromides,freely soluble in alcohol,ether,chcl3,ccl4,cs2 |
| Show More |
Safety and Handling
| Hazard Category | GHS Signal Word: Danger |
| Hazard Statement | GHS H Statement Causes severe skin burns and eye damage. Fatal if inhaled. Very toxic to aquatic life. |
| Precautionary Statement | GHS P Statement IF SWALLOWED: rinse mouth. Do NOT induce vomiting. Wear protective gloves/protective clothing/eye protection/face protection. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses,if pre |
| EINECSNumber | 231-778-1 |
RUO – Research Use Only