missing translation for 'onlineSavingsMsg'
Learn More

Bromine - 1,4-Dioxane Complex 95.0+%, TCI America™

Catalog No. p-7119738
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B27195G 5 g
B271925G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B27195G Supplier TCI America Supplier No. B27195G

Chemical Identifiers

CAS 15481-39-7
Molecular Formula C4H8BrO2
Molecular Weight (g/mol) 168.01
InChI Key GSQACUNMFGRAKY-UHFFFAOYSA-N
Synonym p-dioxane, compd. with bromine 1:1
PubChem CID 57348859
IUPAC Name bromine;1,4-dioxane
SMILES C1COCCO1.[Br]

Specifications

Melting Point 68°C
Quantity 5 g
Physical Form Crystal-Powder at 20°C
UN Number 1325
Boiling Point 95°C
Formula Weight 247.91
Percent Purity ≥95.0% (T)
Chemical Name or Material Bromine - 1,4-Dioxane Complex
TSCA No
Recommended Storage Freezer