missing translation for 'onlineSavingsMsg'
Learn More

Bromide Standard, 500 ppm Br-, Ricca Chemical

Catalog No. R1178000120
Change view
Click to view available options
Quantity:
120 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
R1178000120 120 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. R1178000120 Supplier Ricca Chemical Company Supplier No. R1178000120A

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 7647-15-6 , 7732-18-5
Molecular Formula BrNa
Molecular Weight (g/mol) 102.89
MDL Number MFCD00003475
InChI Key JHJLBTNAGRQEKS-UHFFFAOYSA-M
PubChem CID 253881
ChEBI CHEBI:63004
IUPAC Name sodium bromide
SMILES [Na+].[Br-]

Specifications

Packaging Natural Poly Bottle
CAS Min % 98.0, 0.06
CAS Max % 100.0, 0.07
Equivalence 1 mL = 0.500 mg Br
Physical Form Liquid
Color Colorless
pH 6.5 to 8
Melting Point 0.0°C
Boiling Point 100°C
For Use With (Equipment) AA, ICP, ICP/MS, IC, XRF, and Other Analytical Instrumentation
Solubility Miscible with water
Solution Type Calibration Standard
Type Analytical Standard
Quantity 120 mL
Grade Laboratory
Chemical Name or Material Bromide Standard
Show More Show Less