missing translation for 'onlineSavingsMsg'
Learn More

Bromhexine Hydrochloride 98.0+%, TCI America™

Catalog No. B405425G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B405425G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B405425G Supplier TCI America Supplier No. B405425G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 611-75-6
Molecular Formula C14H21Br2ClN2
Molecular Weight (g/mol) 412.59
MDL Number MFCD00056626
InChI Key SXIVGYTWSVJOLM-UHFFFAOYNA-N
Synonym bromhexine hydrochloride, bromhexine hcl, auxit, bisolvon, broncokin, viscolyt, bromohexine hydrochloride, bromhexine chloride, quentan, unii-yc2zom3z8v
PubChem CID 5702220
ChEBI CHEBI:31303
IUPAC Name hydrogen 2-(1-amino-1-cyclohexylethyl)-4,6-dibromoaniline chloride
SMILES [H+].[Cl-].CC(N)(C1CCCCC1)C1=CC(Br)=CC(Br)=C1N

Specifications

Melting Point 234°C
Color White-Yellow
Quantity 25 g
Formula Weight 412.59
Percent Purity ≥98.0% (HPLC,N)
Physical Form Crystalline Powder
Chemical Name or Material Bromhexine Hydrochloride
EINECSNumber (9)-0516
RTECSNumber XS9950000
TSCA No