missing translation for 'onlineSavingsMsg'
Learn More

Bromate-Bromide Solution, 0.500 N (N/2), 0.250 M as Br2, Ricca Chemical

Catalog No. R11720004F
Change view
Click to view available options
Quantity:
500 mL
1 L
4 L
Packaging:
Amber Glass Bottle
Cubitainer
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
R11720004F 4 L Cubitainer
117216 500 mL Amber Glass Bottle
117232 1 L Amber Glass Bottle
11721 4 L Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. R11720004F Supplier Ricca Chemical Company Supplier No. R11720004F

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Specifications

Chemical Name or Material Bromate-Bromide Solution
Color Colorless
CAS 7758-02-3, 7758-01-2
CAS Min % 92.0, 4.7, 1.28
CAS Max % 96.0, 4.89, 1.33
Molecular Formula BrK
MDL Number MFCD00011358
Synonym potassium bromide, bromide salt of potassium, kalii bromidum, bromure de potassium, caswell no. 684, tripotassium tribromide, potassium bromide k3br3, unii-osd78555zm, ccris 6095
InChI Key IOLCXVTUBQKXJR-UHFFFAOYSA-M
SMILES [K+].[Br-]
IUPAC Name potassium bromide
Molecular Weight (g/mol) 119.00
PubChem CID 253877
ChEBI CHEBI:32030
Grade Laboratory
Packaging Cubitainer
Quantity 4 L
Physical Form Liquid
Concentration or Composition Notes 0.500Normal,0.250Molar as Br
Show More Show Less