missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Bromate-Bromide Solution, 0.0500 N (N/20), Ricca Chemical
Specifications
Specifications
| Chemical Name or Material | Bromate-Bromide Solution |
| Color | Colorless |
| CAS | 7732-18-5, 7758-01-2, 7758-01-2 |
| CAS Min % | 97.0, 0.97, 0.13 |
| CAS Max % | 100.0, 1.01, 0.14 |
| Synonym | dihydrogen oxide, dihydrogen monoxide |
| Grade | Laboratory |
| Packaging | Amber Glass Bottle |
| Quantity | 1 L |
| Physical Form | Liquid |
| Show More |