missing translation for 'onlineSavingsMsg'
Learn More

Bromate-Bromide Solution, 0.0250 N (N/40), Ricca Chemical

Supplier:  Ricca Chemical Company 11661

 View more versions of this product

Catalog No. R11661


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

Bromate-Bromide Solution
7732-18-5 , 7758-01-2 , 7758-01-2
100.0,1.01,0.07
Laboratory
4 L
0.0250Normal
Colorless
97.0,0.97,0.07
dihydrogen oxide, dihydrogen monoxide
Amber Glass Bottle
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions