missing translation for 'onlineSavingsMsg'
Learn More

Bromate-Bromide Solution, 0.0250 N (N/40), Ricca Chemical

$658.67 - $658.67

Specifications

Chemical Name or Material Bromate-Bromide Solution
Color Colorless
CAS 7732-18-5 , 7758-01-2 , 7758-01-2
CAS Min % 97.0,0.97,0.07
CAS Max % 100.0,1.01,0.07
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
R11661
View Documents Promotion Details
Ricca Chemical Company
11661
4 L
Each for $658.67
Only null left
 
Description

Description

Specifications

Specifications

Bromate-Bromide Solution
7732-18-5 , 7758-01-2 , 7758-01-2
100.0,1.01,0.07
Laboratory
Liquid
Colorless
97.0,0.97,0.07
dihydrogen oxide, dihydrogen monoxide
Amber Glass Bottle
0.0250Normal
Videos
SDS
Documents

Documents