missing translation for 'onlineSavingsMsg'
Learn More

Bromate-Bromide, Certified, 0.500N ±0.005N (0.083M), LabChem™

Catalog No. LC119704
Change view
Click to view available options
Quantity:
500 mL
4 L
Packaging:
Amber Glass
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
LC119704 4 L Amber Glass
LC119701 500 mL Amber Glass
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. LC119704 Supplier LabChem Supplier No. LC119704

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Specifications

Chemical Name or Material Bromate-Bromide
CAS Max % 93.1, 5.5, 1.4
CAS 7758-01-2
Color Colorless
Packaging Amber Glass
Linear Formula KBrO3
Quantity 4 L
Synonym water, dihydrogen oxide, distilled water, purified water, water vapor, sterile water, water, purified, water, deionized, water, mineral, dihydrogen monoxide
Solubility Information Soluble in water
Formula Weight 167
Grade Certified
Concentration or Composition (by Analyte or Components) 0.500N ±0.005N (0.083M)
Physical Form Liquid
Show More Show Less