missing translation for 'onlineSavingsMsg'
Learn More

Brivudine, 98%

Numéro de catalogue. aah2785303
Modifier l'affichage
Click to view available options
Quantity:
50 mg
250 mg
1 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantity
AAH2785303 1 g
AAH27853MF 50 mg
AAH27853MD 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Numéro de catalogue. AAH2785303 Fournisseur Thermo Scientific Chemicals Code fournisseur H2785303
Il en reste null

CAS: 69304-47-8 | C11H13BrN2O5 | 333.14 g/mol

It is used as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used as pharmaceutical intermediate.

Solubility
Soluble in water and methanol.

Notes
Store in cool dry conditions. Incompatible with oxidizing agents.

Identifiants chimiques

CAS 69304-47-8
Molecular Formula C11H13BrN2O5
Molecular Weight (g/mol) 333.14
MDL Number MFCD00058585
InChI Key ODZBBRURCPAEIQ-PIXDULNESA-N
Synonym brivudine, bvdu, brivudin, helpin, e-5-2-bromovinyl-2'-deoxyuridine, bromovinyldeoxyuridine, zostex, brivudine inn, brivudinum inn-latin, brivudina inn-spanish
PubChem CID 446727
IUPAC Name 5-[(E)-2-bromoethenyl]-1-[(2R,4S,5R)-4-hydroxy-5-(hydroxymethyl)oxolan-2-yl]pyrimidine-2,4-dione
SMILES OC[C@H]1O[C@H](C[C@@H]1O)N1C=C(\C=C\Br)C(=O)NC1=O

Spécifications

Melting Point ∽165°C (decomposition)
Quantity 1 g
Solubility Information Soluble in water and methanol.
Optical Rotation +10° (c=1 in Methanol)
Formula Weight 333.14
Percent Purity 98%
Chemical Name or Material (E)-5-(2-Bromovinyl)-2'-deoxyuridine
RTECSNumber YU7355000
TSCA No
Recommended Storage Keep cold

RUO – Research Use Only