missing translation for 'onlineSavingsMsg'
Learn More

Brinzolamide 98.0+%, TCI America™

Catalog No. B425825MG
Change view
Click to view available options
Quantity:
25 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B425825MG 25 mg
B4258100MG 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B425825MG Supplier TCI America Supplier No. B425825MG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 138890-62-7
Molecular Formula C12H21N3O5S3
Molecular Weight (g/mol) 383.496
MDL Number MFCD08067749
InChI Key HCRKCZRJWPKOAR-JTQLQIEISA-N
Synonym (R)-4-(Ethylamino)-2-(3-methoxypropyl)-3,4-dihydro-2H-thieno[3,2-e][1,2]thiazine-6-sulfonamide 1,1-Dioxide
PubChem CID 68844
ChEBI CHEBI:41212
IUPAC Name (4R)-4-(ethylamino)-2-(3-methoxypropyl)-1,1-dioxo-3,4-dihydrothieno[3,2-e]thiazine-6-sulfonamide
SMILES CCNC1CN(S(=O)(=O)C2=C1C=C(S2)S(=O)(=O)N)CCCOC

Specifications

Melting Point 132°C
Color White-Yellow
Quantity 25 mg
Formula Weight 383.50
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material Brinzolamide
RTECSNumber XJ9095055
TSCA No
Recommended Storage Freezer