missing translation for 'onlineSavingsMsg'
Learn More

Metrohm Accessories and Replacement Parts for Models 756 and 831 KF Coulometers

Catalog No. BR20928905
Change view
Click to view available options
Temperature (Metric) Range:
-20°C to +70°C
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Temperature (Metric) Range
BR20928905 -20°C to +70°C
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BR20928905 Supplier Metrohm Supplier No. 60341100

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Specifications

Type Electrode
Temperature (English) Range -4°F to 158°F
Temperature (Metric) Range -20°C to +70°C