missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Brilliant Green, Reagent Grade, LabChem™
Specifications
Specifications
| Melting Point | 210°C |
| CAS Max % | 100 |
| Linear Formula | C27H33N2·HO4S |
| Solubility Information | Soluble in water |
| Identification | Passes Test |
| Formula Weight | 482.65 |
| Grade | Reagent |
| Packaging | Amber Glass |
| Decomposition Information | Nitrogen oxides; Carbon monoxide; Carbon dioxide; Sulfur compounds |
| Color | Gray/Green |
| Show More |
Safety and Handling
| Hazard Category | Warning |
| Hazard Statement | GHS H Statement Harmful if swallowed. Causes serious eye irritation. |
| Precautionary Statement | GHS P Statement Wear protective gloves, eye protection. Wash exposed skin thoroughly after handling. Do not eat, drink or smoke when using this product. If swallowed: Rinse mouth. Call a poison center/doctor if you feel unwell. If in eyes: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. If eye irritation persists: Get medical advice/attention. Dispose of contents/container to comply with local, state and federal regulations. |
| Recommended Storage | Room Temperature |