missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Brilliant Green, pure, high purity biological stain

Catalog No. AC229605000
Change view
Click to view available options
Quantity:
100 g
500 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC229605000 500 g Glass bottle
AC229601000 100 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC229605000 Supplier Thermo Scientific Chemicals Supplier No. 229605000
Only null left

CAS: 633-03-4 | C27H34N2O4S | 482.639 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 633-03-4
Molecular Formula C27H34N2O4S
Molecular Weight (g/mol) 482.639
MDL Number MFCD00011880
InChI Key NNBFNNNWANBMTI-UHFFFAOYSA-M
Synonym Basic Green 1, C.I. 42040, Ethyl Green
PubChem CID 12449
IUPAC Name [4-[[4-(diethylamino)phenyl]-phenylmethylidene]cyclohexa-2,5-dien-1-ylidene]-diethylazanium;hydrogen sulfate
SMILES CCN(CC)C1=CC=C(C=C1)C(=C2C=CC(=[N+](CC)CC)C=C2)C3=CC=CC=C3.OS(=O)(=O)[O-]

Specifications

Absorbance 1875 (at lambda min.)
Melting Point 180.0°C
Absorption Ratio 1 to 1.2
Beilstein 13, 746
Merck Index 15, 1380
Solubility Information Solubility in water: 100g/L water (25°C). Other solubilities: soluble in alcohol
Infrared Spectrum Authentic
Lambda Max 625 to 632nm (in 50:50 IMS:water)
Formula Weight 482.64
Loss on Drying 10% max. (105°C)
Grade Pure
Packaging Glass bottle
Color Green
Quantity 500 g
Physical Form Crystalline Powder or Crystals
Chemical Name or Material Brilliant Green, High purity biological stain
Show More Show Less
Hazard Category Warning
Hazard Statement GHS H Statement:
Harmful if swallowed.
Causes serious eye irritation.
Precautionary Statement GHS P Statement:
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

RUO – Research Use Only