missing translation for 'onlineSavingsMsg'
Learn More

Brilliant Blue FCF, ≥97% (HPLC), MilliporeSigma™ Supelco™

Catalog No. 111014100
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111014100 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111014100 Supplier Merck Emd Millipore Supplier No. 80717100MG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Brilliant blue FCF has been used as a test compound in validating the lowest observed adverse effect level (LOAEL) to inflict developmental toxicity upon exposure to common excipients.

Featured Industries: Cleaning Products, Cosmetics, Food and Beverages and Personal Care

Specifications

CAS 3844-45-9
Physical Form Neat
Linear Formula C37H34 Na2N2O9S3
MDL Number MFCD00012141
Melting Point 283°C
Synonym Erioglaucine disodium salt; E133; Acid Blue 9; Alphazurine FG; FD&C;BLUE No. 1
Type Analytical Standard
Quantity 100 mg
Formula Weight 792.85
Percent Purity ≥97% (HPLC)
Grade Analytical Standard
Show More Show Less