missing translation for 'onlineSavingsMsg'
Learn More

Brij(R) S100, Spectrum™ Chemical

Catalog No. 18601069
Change view
Click to view available options
Quantity:
25 g
500 g
2.5 kg
Packaging:
Amber Glass Bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18601069 25 g Amber Glass Bottle
18601070 500 g Amber Glass Bottle
18601071 2.5 kg Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 18601069 Supplier Spectrum Chemical Mfg Cor Supplier No. B159325GM
Only null left

B1593, 9005-00-9

Spectrum™ Chemical Brij S100 , Polyethylene (100) Stearyl Ether, functions as an aqueous dispersant and oil/water emulsifier.

Chemical Identifiers

CAS 9005-00-9
Molecular Formula C20H41O2
Molecular Weight (g/mol) 313.55
InChI Key PSEGVHKUPXNGGJ-UHFFFAOYSA-N
IUPAC Name [2-(octadecyloxy)ethyl]oxidanyl
SMILES CCCCCCCCCCCCCCCCCCOCC[O]

Specifications

CAS Max % 100%
Quantity 25 g
Packaging Amber Glass Bottle
Grade Ungraded
Chemical Name or Material Brij 700