missing translation for 'onlineSavingsMsg'
Learn More

Brij(R) 35, 30% (w/v) Solution, Spectrum™ Chemical

Catalog No. 18609198
Change view
Click to view available options
Quantity:
1 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
18609198 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 18609198 Supplier Spectrum Chemical Mfg Cor Supplier No. BR1101LT

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Specifications

Chemical Name or Material Brij 35
CAS 9002-92-0, 7732-18-5
Concentration 30%
Quantity 1 L
Packaging Poly Bottle
Grade ACS

CAUTION: For manufacturing or laboratory use only. Not for food or drug use. Read and understand the label and Safety Data Sheet (SDS) prior to use.