missing translation for 'onlineSavingsMsg'
Learn More

Brij™ 30

Catalog No. AC216725000
Change view
Click to view available options
Quantity:
500 g
2.5 kg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC216725000 500 g
AC216720025 2.5 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC216725000 Supplier Thermo Scientific Chemicals Supplier No. 216725000

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 9002-92-0 | C14H30O2 | 230.39 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 9002-92-0
Molecular Formula C14H30O2
Molecular Weight (g/mol) 230.39
MDL Number MFCD00043063
InChI Key SFNALCNOMXIBKG-UHFFFAOYSA-N
Synonym Polyoxyethylene(4)lauryl ether
IUPAC Name 2-(dodecyloxy)ethan-1-ol
SMILES CCCCCCCCCCCCOCCO

Specifications

Color Colorless to Yellow
Acid Value 2mg KOH/g max.
Physical Form Liquid
Melting Point 14.0°C
Density 0.9500g/mL
Quantity 500 g
Flash Point >149°C
Hydroxyl Value 145 to 165mg KOH/g
Infrared Spectrum Authentic
Packaging Glass bottle
Refractive Index 1.4510 to 1.4530
Specific Gravity 0.95
Viscosity 30 mPa.s (25°C)
Linear Formula C12H25(OCH2CH2)4OH
Water 1% max.
Fieser 01,892
Merck Index 15, 7675
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in lower alcohols, propylene glycol, insoluble in mineral oil, cottonseed oil
Formula Weight 362.54
Chemical Name or Material Brij™ 30
Show More Show Less
Hazard Category Danger
Hazard Statement GHS H Statement
Causes serious eye damage.
Precautionary Statement GHS P Statement
Wear eye protection/face protection. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. Immediately call a POISON CENTER or doctor/physician.
RTECSNumber MD0875000
TSCA TSCA

RUO – Research Use Only