missing translation for 'onlineSavingsMsg'
Learn More

BRANDTECH™ BRAND™ Accessories for PLT™ (Pipette Leak Testing) Units

Catalog No. 14726831
Change view
Click to view available options
Type:
Multichannel Pipet Adapter
Replacement Filters
Single Channel Pipette Adapter
Universal Power Supply
For Use With (Application):
Testing with or without Tips
Testing with Tips
Testing without Tips
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Type For Use With (Application)
14726831 Single Channel Pipette Adapter Testing with Tips
13688238 Multichannel Pipet Adapter Testing with or without Tips
13688239 Replacement Filters -
14726832 Single Channel Pipette Adapter Testing without Tips
14726833 Universal Power Supply -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 14726831 Supplier BRANDTECH™ BRAND™ Supplier No. 703975

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

For use with BRAND™ PLT™ unit. These accessories are designed for improved process reliability in identifying even the smallest leaks in your air displacement pipette.

Available accessories include:

  • Part No. 703978: Polyethylene filters and cap for pipette adapters, includes ten filters and one cap.
  • Part No. 703977: Four-channel pipette adapter, for testing with and without tips, includes four plugs.
  • Part No. 703975: Single-channel pipette adapter for testing with tips, includes one plug.
  • Part No. 703976: Single-channel pipette adapter for testing without tips, includes one plug.
  • Part No. 703979: Universal AC Adapter, 100-240V, 50/60Hz.

Specifications

Compatible Tips All tips
Description Single Channel Pipette Adapter, For testing with tips, Includes plugs
Autoclavable Not Autoclavable
Color Transparent/White
Disposable Reusable
For Use With All Pipettes with Tips Mounted
Material Silicone
Number of Channels 1
Sterility Non-sterile
Type Single Channel Pipette Adapter
For Use With (Application) Testing with Tips
For Use With (Equipment) PLT Pipette Leak Testing Unit
Includes Plug
Show More Show Less