missing translation for 'onlineSavingsMsg'
Learn More

BRANDTECH™ BRAND™ Cuvette Caps

Catalog No. 14377007
Change view
Click to view available options
Color:
Blue
Green
Yellow
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
14377007 Green
13878125 Blue
14377006 Yellow
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 14377007 Supplier BRANDTECH™ BRAND™ Supplier No. 759242

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Provides tight seal, allows storage of samples to -20°C.

  • Round cuvette caps
  • Fits ultra-micro cuvettes
  • Provide tight seal
  • Allows storage of samples to -20°

Specifications

Type Cuvette Cap
Color Green
Disposable Single-use
Material Polyethylene
Quantity 100 Pack
For Use With (Equipment) Ultramicro UV Cuvette
Shape Round