Learn More
BRANDTECH™ BRAND™ BRANDplates™ pureGrade™ 96-Well Microplates
Constructed of polystyrene with nontreated surface. BRANDplates™ pureGrade™ 96-Well Microplates are noncytotoxic, and free from endotoxins, DNase, DNA, and RNase. Medium binding, nonsterile microplates are suited for homogenous assays, screening, and storage with gray alphanumeric embossing on white and black solid plates for easy well identification.
 View more versions of this product
Catalog No. 13882204
Only null left
Description
Description
  • Polystyrene surface
  • Medium binding, nonsterile
  • Standard plate for most applications
  • Gray alphanumeric embossing on white and black solid plates for easy well identification
  • Strip plates available
  • Standard plates available in U-, V-, F-, C-shapes
  • Transparent bottoms available in F- shape
  • Available in clear, white or black
  • Free from endotoxins, DNase, DNA, RNase; noncytotoxic
  • Suited for homogenous assays, screening, and storage.
Specifications
Chemical Identifiers
Includes Barcode
Standard
Polystyrene
Non-sterile
Homogenous assays, screenings and storage
ANSI, SLAS
8 x 12
96
Yes
Yes
Specifications
Includes Barcode
Standard
8 x 12
White
Polystyrene
96
Non-treated
Yes
100/Cs.
ANSI, SLAS
Without Lid
Medium Binding
Standard
330 μL
Non-sterile
Round Bottom (U-well)
Homogenous assays, screenings and storage
Yes
Product Suggestions
Product Suggestions