Learn More
BRANDTECH™ BRAND™ Plastic Spectrophotometry Cuvettes
BRAND™ Plastic Spectrophotometry Cuvettes with mold-cavity matched for the lowest variability in extinction coefficient.
Supplier:  BRANDTECH™ BRAND™ 759076D
 View more versions of this product
Catalog No. 14377013
Only null left
Description
Description
BRAND spectrophotometry cuvettes. Mold-cavity matched for the lowest variability in extinction coefficient.
  • Mold cavity sorted spectrophotometry cuvettes with 2 clear windows
Specifications
Chemical Identifiers
Non-sterile
Clear
Arrow mark to indicate direction of transmission and reduce variation
10 mm
Spectrophotometers, Photometers
Spectrophotometry
Rectangle
Spectrophotometer Cuvette
Single-use
Life Science Quality
45 mm
12.5 mm
10 mm
Specifications
Non-sterile
Semi-micro
Single-use
Polystyrene
Life Science Quality
10 mm
45 mm
12.5 mm
10 mm
Spectrophotometer Cuvette
Clear
3 mL
Arrow mark to indicate direction of transmission and reduce variation
100 Pk.
340 nm
Spectrophotometers, Photometers
Spectrophotometry
Rectangle
Product Suggestions
Product Suggestions