missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Brady™ BradyPrinter S3700 Multicolor and Cut Sign and Label Printer with Lean/5S Label Materials Kit
Get everything you need to easily create a visual workplace with this Lean/5S label kit. From materials to software, this kit has you covered.
Supplier: Brady™ S3700KITLEAN
Description
- Printer optimized for 1-, 2- or 4-color panel ribbons for multicolor made easy
- No calibrating, no threading, no waste (single-color ribbon only in kit)
- Create shapes, arrows, letters and more with the built-in XY plotter cutter that slices straight lines, smoother curves and tighter connection points
- Get your print jobs done right in less time with 20-second ribbon and material changeovers plus up to 5 in./sec. print speed
- Empower any user to print how they choose with time-saving onboard templates, intuitive PC labeling software and a mobile app
- Repositionable Magnetic Labels and printable ToughStripe™ floor tape are perfect for bin labeling, area marking and other Lean/5S applications
- Durable, low-shrink All Weather Permanent Adhesive Vinyl Label Tape with ultra-aggressive adhesive is ideal for just about any industrial surface
- Waste less, print more with print ribbons that last up to 60% longer by operating independently of label roll
Spécifications
| Label Printer Kit | |
| Color LCD Touchscreen | |
| Plastic | |
| 14.25 in. | |
| 361.95 mm | |
| 11 in. | |
| 14.25 x 19 x 11 in. | |
| Non-sterile | |
| S3700 Safety and Facility Printer, Brady Workstation Safety and Facility ID Software, Cleaning Kit, Mouse, Power Cord, Quick Start Guide, USB Flash Drive, 4 in. White and Yellow Vinyl Continuous Labels, 2.25 in. White and Yellow Continuous Vinyl Labels, 3.85 x 2.25 in. White Vinyl Label, 2.5 in. Continuous Magnetic Polyester Label, 4.25 in. Continuous Magnetic Polyester Label, 2.25 in. Continuous Yellow Polyester Label, Black Ribbon Cartridge, White Ribbon Cartridge, Red Ribbon Cartridge, Blue Ribbon Cartridge, 4-color Ribbon Cartridge | |
| 512 MB | |
| 5 in./sec. | |
| 59°F to 104°F | |
| Asset Tracking, Cold Temperature Labeling, Equipment Labeling, Electrical Labeling, General Identification, Hazard Communication Identification, Lean and 5S Labeling, Food and Beverage Process Labeling, Healthcare and Hospital Labeling, Barcode Labeling | |
| AC Adaptor | |
| QWERTY (US) |
| Blue, Gray | |
| Brady Labels | |
| Thermal Transfer | |
| 19 in. | |
| 482.6 mm | |
| 279.4 mm | |
| Not Autoclavable | |
| Multicolor | |
| Indonesian, Bulgarian, Chinese (Simplified), Chinese (Traditional), Croatian, Czech, Danish, Dutch, English, Estonian, Finnish, French, German, Hungarian, Italian, Japanese, Korean, Norwegian, Polish, Portuguese, Romanian, Russian, Slovak, Slovenian, Spanish, Swedish, Thai, Turkish, Vietnamese | |
| S3700 | |
| 300 dpi | |
| 15°C to 40°C | |
| 5 Years | |
| 110/240 V |