missing translation for 'onlineSavingsMsg'
Learn More

Brady™ BradyPrinter i5300 Industrial Label Printer 600 dpi WiFi PWID Software Suite

Catalog No. 17607630
Change view
Click to view available options
Color:
Blue, Gray
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
17607630 Blue, Gray
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 17607630 Supplier Brady™ Supplier No. 151294

Get fast and efficient precision printing for high-mix / high-volume labeling. Printer and software allow for quick label set up and no-waste material changeovers.

Get fast and efficient precision printing for high-mix/high-volume labeling.

  • Printer and software allow for quick label set up and no-waste material changeovers.
  • 600 dpi, prints small fonts with precision.
  • Requires no calibration and no heat and speed setting adjustments.
  • 20-second material changeovers, 0.20 to 4 in. label widths.
  • Pauses, resumes and stores print jobs on printer in an adjustable queue with overwrite protection.
  • 7 in. color touchscreen and built-in auto cutter.
  • Solid construction with an all-metal frame.
  • 1-click label part recognition and setup with Brady Workstation Product and Wire ID Software Suite.
  • Intuitive and easy to operate.

Specifications

Product Type Industrial Label Printer
Color Blue, Gray
Display Type Color LCD Touchscreen
For Use With (Equipment) B30 Series Thermal Transfer Tapes and Ribbons
Material Plastic
Print Technology Direct Thermal, Thermal Transfer
Length (English) 15.8 in.
Width (English) 11.2 in.
Autoclavable Not Autoclavable
Sterility Non-sterile