missing translation for 'onlineSavingsMsg'
Learn More

Brady™ BradyPrinter i3300 Rotating Self-laminating Vinyl Wire Labels

Identify your wires from any angle with B-427 Rotating Vinyl Wire Labels. Simply print your labels and then peel, fold, and wrap – they’ll rotate freely for easy ID on even closed network connections.

$159.02 - $458.65

Specifications

Product Type Wire Labels
Color White/Translucent
For Use With (Equipment) BradyPrinter i3300
Material Self-laminating Vinyl
Adhesive Type Permanent
View More Specs

Products 7
Catalog Number Mfr. No. Width (English) Height (English) Price Quantity & Availability  
Catalog Number Mfr. No. Width (English) Height (English) Price Quantity & Availability  
19129513
View Documents Promotion Details
Brady™
B33RO292427
0.5 in. 1.75 in.
Each for $214.41
Only null left
 
19129514
View Documents Promotion Details
Brady™
B33RO293427
0.5 in. 2.75 in.
Each for $214.41
Only null left
 
19129515
View Documents Promotion Details
Brady™
B33RO294427
1 in. 1.75 in.
Each for $275.44
Only null left
 
19129516
View Documents Promotion Details
Brady™
B33RO295427
1 in. 2.75 in.
Each for $312.37
Only null left
 
19129517
View Documents Promotion Details
Brady™
B33RO296427
1 in. 3.125 in.
Each for $458.65
Only null left
 
19129518
View Documents Promotion Details
Brady™
B33RO297427
1 in. 1.2 in.
Each for $159.02
Only null left
 
19129519
View Documents Promotion Details
Brady™
B33RO299427
2 in. 3.125 in.
Each for $399.00
Only null left
 
Description

Description

  • Rotates so you can identify your labels from any angle
  • Simply wrap and apply – there is no adhesive residue or label parts to cling to your wires
  • Spins freely without anchors so these labels won't damage your Cat 5/6 and fiber optic cables
Specifications

Specifications

Wire Labels
BradyPrinter i3300
Permanent
Rectangle
Wire and Cable Labeling
White/Translucent
Self-laminating Vinyl
Matte
-94°F to +158°F
Videos
SDS
Documents

Documents